Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1110362
Encompass_Preferred
Fabric heating mantle for 400ml griffin beaker to 400 degrees C maximum internal operating temperature, 240W, 115V, CSA
Catalog No. NC1110362 Supplier Glas Col LLC Supplier No. 100A O608
Fabric heating mantle for 400ml griffin beaker to 400 degrees C maximum internal operating temperature, 240W, 115V, CSA