Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1811609
Encompass_Preferred
Aluminum housed mantle for 1 gallon can, to 400 degrees C maximum internal operating temperature, 750W, 115V.
Catalog No. NC1811609 Supplier Glas Col LLC Supplier No. 100D DH098BC
Aluminum housed mantle for 1 gallon can, to 400 degrees C maximum internal operating temperature, 750W, 115V.