Learn More
V&P Scientific Inc HEATER/CHILLER, Recirculating Fluid Block, for Deep or Standard Well Format
Catalog No. NC2250229
Encompass
HEATER/CHILLER, Recirculating Fluid Block, for Deep or Standard Well Format, Special Design to Minimize Eddy Currents when used on a Tumble Stirrer, VP 581B-MICA SLAS Feature on Bottom, Includes Screw Down Lid, with Low Temperature Fittings, Designed to Operate at -70°C
This item is not returnable. View return policy
Catalog No. NC2250229 Supplier V&P Scientific Inc Supplier No. VP 744BMBLT70
Add to Cart
Add to Cart
Request a Quote
HEATER/CHILLER, Recirculating Fluid Block, for Deep or Standard Well Format, Special Design to Minimize Eddy Currents when used on a Tumble Stirrer, VP 581B-MICA SLAS Feature on Bottom, Includes Screw Down Lid, with Low Temperature Fittings, Designed to Operate at -70°C