Learn More
MSC Heatstar Portable Radiant Heater, Natural Gas, 125,000 BTU, CSA Certified
Catalog No. 5000507708
Encompass
This portable radiant heater is designed for efficient heating in various environments. Powered by natural gas, it delivers a robust 125,000 BTU, capable of heating areas up to 3000 square feet.
  • Portable design for easy movement and placement.
  • Operates on natural gas for convenient fuel access.
  • Provides 125,000 BTU maximum heating capacity.
  • Heats up to 3000 square feet.
  • Complies with CSA safety standards.
  • Compact dimensions of 16-1/4 in wide, 16-1/4 in deep, and 35 in high.
Catalog No. 50-005-07708 Supplier MSC Supplier No. 93178879
This portable radiant heater is designed for efficient heating in various environments. Powered by natural gas, it delivers a robust 125,000 BTU, capable of heating areas up to 3000 square feet.
  • Portable design for easy movement and placement.
  • Operates on natural gas for convenient fuel access.
  • Provides 125,000 BTU maximum heating capacity.
  • Heats up to 3000 square feet.
  • Complies with CSA safety standards.
  • Compact dimensions of 16-1/4 in wide, 16-1/4 in deep, and 35 in high.