Learn More
Ironclad Performance Wear Heatworx Heavy Duty LARGE
Catalog No. NC0248236
Encompass_Preferred
This item must be ordered in multiples of 12 pair per supplier requirements.
Ironclad HEATWORX HEAVY DUTY Work Gloves; , Molded Silicone Palm Protects Heat up to 600°, DuPont™ Kevlar Back of Hand, Slip On Cuff, Kevlar™ Liner & Stitching, Silicone Coated FR Knuckle, Reinforced Thumb Saddle, Sized S - XXL (1 pair) - HW6X-04-L
This item is not returnable. View return policy
Catalog No. NC0248236 Supplier Ironclad Performance Wear Supplier No. HW6X04L
This item must be ordered in multiples of 12 pair per supplier requirements.
Ironclad HEATWORX HEAVY DUTY Work Gloves; , Molded Silicone Palm Protects Heat up to 600°, DuPont™ Kevlar Back of Hand, Slip On Cuff, Kevlar™ Liner & Stitching, Silicone Coated FR Knuckle, Reinforced Thumb Saddle, Sized S - XXL (1 pair) - HW6X-04-L