Learn More
Cayman Chemical Heavy ChaInhumn Monclo 100ug
SDP
Catalog No. 502946338
Encompass_Preferred
Immunogen: Human IgG • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgG1, IgG2, IgG3, IgG4; (-) Human IgM, IgA, IgD, IgE; (-) Goat, mouse, rat IgG • Applications: ELISA, ICC, IHC
Catalog No. 50-294-6338 Supplier Cayman Chemical Supplier No. 32107100 UG
Immunogen: Human IgG • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgG1, IgG2, IgG3, IgG4; (-) Human IgM, IgA, IgD, IgE; (-) Goat, mouse, rat IgG • Applications: ELISA, ICC, IHC