Learn More
MSC ZEP Liquid Carpet Cleaner, Lemongrass Scent, Heavy-Duty, 5 gal
Catalog No. 502815969
Encompass
This clear, liquid carpet cleaner is formulated for heavy-duty applications. It is designed to rapidly penetrate and remove tenacious soil, grease, organic stains, and odors from carpets.
  • Pleasant lemongrass scent
  • Special heavy-duty formulation
  • Rapidly penetrates and removes tenacious soil and grease
  • Removes organic stains and odors
  • Concentrated for effective cleaning
  • Suitable for carpet care
  • Has a pH range of 8.5-9.5
Catalog No. 50-281-5969 Supplier MSC Supplier No. 10830123
This clear, liquid carpet cleaner is formulated for heavy-duty applications. It is designed to rapidly penetrate and remove tenacious soil, grease, organic stains, and odors from carpets.
  • Pleasant lemongrass scent
  • Special heavy-duty formulation
  • Rapidly penetrates and removes tenacious soil and grease
  • Removes organic stains and odors
  • Concentrated for effective cleaning
  • Suitable for carpet care
  • Has a pH range of 8.5-9.5