Learn More
Chemglass Life Sciences Heavy Duty Clamp Holder, Fits 3/4" Support Rod, Silver Epoxy Coated, fits ChemRxnHub
SDP
Catalog No. 501531411
Encompass_Preferred
Heavy Duty Clamp Holder, Fits 3/4" Support Rod. Heavy duty epoxy coated alloy clamp has high tensile strength and corrosion resistance. Can be used to support stirrers, temperature monitors and condensers. Large hand knobs allow easy adjusment and secure clamping power. Useful wherever clamping at 90 is required.
Catalog No. 50-153-1411 Supplier Chemglass Life Sciences Supplier No. CG2040C20
Heavy Duty Clamp Holder, Fits 3/4" Support Rod. Heavy duty epoxy coated alloy clamp has high tensile strength and corrosion resistance. Can be used to support stirrers, temperature monitors and condensers. Large hand knobs allow easy adjusment and secure clamping power. Useful wherever clamping at 90 is required.