Learn More
Grainger GRAINGER APPROVED Heavy Duty Clamp, 304 Stainless Steel
Catalog No. NC2592586
Encompass_Preferred
The Grainger Approved Heavy Duty Clamp is made from 304 stainless steel with a satin finish. It has a throat depth of 4" and can exert up to 750 lbs of clamping force. The clamp has a malleable iron handle that provides leverage for tightening. It is designed for industrial use and rated for indoor and outdoor applications. The clamp opens to fit materials up to 3-3/4" thick and has a lightweight design weighing just 1.4 lbs.
Catalog No. NC2592586 Supplier Grainger Supplier No. 11P806
The Grainger Approved Heavy Duty Clamp is made from 304 stainless steel with a satin finish. It has a throat depth of 4" and can exert up to 750 lbs of clamping force. The clamp has a malleable iron handle that provides leverage for tightening. It is designed for industrial use and rated for indoor and outdoor applications. The clamp opens to fit materials up to 3-3/4" thick and has a lightweight design weighing just 1.4 lbs.