Learn More
Grainger Heavy Duty Clamp, 304 Stainless
Catalog No. NC2592583
Encompass_Preferred
The Heavy Duty Clamp is made of 304 stainless steel with a working load limit of 5,000 lbs. It has a throat depth of 2-1/2" and an overall length of 7". The clamp has a screw take up to tighten and features a mounting bracket that allows it to be permanently mounted. This heavy duty clamp is designed for industrial applications where high strength and corrosion resistance are needed. The stainless steel construction makes it suitable for use in harsh environments. With its high load capacity, this clamp can be used to securely hold pipes, tubes, and other cylindrical objects in place.
Catalog No. NC2592583 Supplier Grainger Supplier No. 11P805
The Heavy Duty Clamp is made of 304 stainless steel with a working load limit of 5,000 lbs. It has a throat depth of 2-1/2" and an overall length of 7". The clamp has a screw take up to tighten and features a mounting bracket that allows it to be permanently mounted. This heavy duty clamp is designed for industrial applications where high strength and corrosion resistance are needed. The stainless steel construction makes it suitable for use in harsh environments. With its high load capacity, this clamp can be used to securely hold pipes, tubes, and other cylindrical objects in place.