Learn More
MSC ZEP All-Purpose Cleaner, Liquid, Chlorinated, High Foaming, 1 gal
Catalog No. 502819074
Encompass
This heavy-duty, high-foaming, chlorinated liquid all-purpose cleaner is formulated with high foaming detergents, water softeners, chlorine, and alkaline builders. It is designed for robust cleaning applications across various surfaces, effectively tackling dirt, grease, and grime.
  • Rapidly removes dirt, grease, and grime from a variety of surfaces
  • Aids in the removal of protein, blood, and stains, also deodorizes
  • Thick, stable foam clings to vertical surfaces, allowing longer contact time
  • Suitable for use in federally inspected meat and poultry plants
  • Alkaline formula for effective cleaning
  • Bleach-like odor and clear to light yellow color
Catalog No. 50-281-9074 Supplier MSC Supplier No. 17641515
This heavy-duty, high-foaming, chlorinated liquid all-purpose cleaner is formulated with high foaming detergents, water softeners, chlorine, and alkaline builders. It is designed for robust cleaning applications across various surfaces, effectively tackling dirt, grease, and grime.
  • Rapidly removes dirt, grease, and grime from a variety of surfaces
  • Aids in the removal of protein, blood, and stains, also deodorizes
  • Thick, stable foam clings to vertical surfaces, allowing longer contact time
  • Suitable for use in federally inspected meat and poultry plants
  • Alkaline formula for effective cleaning
  • Bleach-like odor and clear to light yellow color