Learn More
MSC ZEP Heavy-Duty Foaming Degreaser, Foaming, Citrus Scent, 16 oz
Catalog No. 502815496
Encompass
Zep 50 Heavy-Duty Foaming Degreaser is a powerful degreaser spray designed to tackle the toughest grease and grime. Its high-performance formula clings to vertical surfaces, extending cleaning time for superior results. It is perfect for engine degreasing, concrete degreasing, and heavy-duty cleaning.
  • Combines solvents and emulsifiers to break down thick grease, oil, and grime effortlessly.
  • Thick, stable foam adheres to vertical surfaces for extended contact and deeper cleaning.
  • Effectively cleans small engines, car and truck engines, and industrial machinery.
  • Removes oil spots and grease stains from concrete floors and shop surfaces.
  • Includes a packet of jet stream nozzle for precise, directional spraying, ensuring targeted cleaning.
Catalog No. 50-281-5496 Supplier MSC Supplier No. 17642919
Zep 50 Heavy-Duty Foaming Degreaser is a powerful degreaser spray designed to tackle the toughest grease and grime. Its high-performance formula clings to vertical surfaces, extending cleaning time for superior results. It is perfect for engine degreasing, concrete degreasing, and heavy-duty cleaning.
  • Combines solvents and emulsifiers to break down thick grease, oil, and grime effortlessly.
  • Thick, stable foam adheres to vertical surfaces for extended contact and deeper cleaning.
  • Effectively cleans small engines, car and truck engines, and industrial machinery.
  • Removes oil spots and grease stains from concrete floors and shop surfaces.
  • Includes a packet of jet stream nozzle for precise, directional spraying, ensuring targeted cleaning.