Learn More
Anton Paar Heavy-duty device with enhanced corrosion resistance, designed for aqua regia and HCl digestions. Software-guided cleaning protocols and an extended maintenance plan ensure reliable performance in demanding applications.
Catalog No. 5000580545
Encompass_Preferred
High temperature and pressure for outstanding digestion resultsSoftware-guided procedures make the instrument easy to useIntegrated cooling system for a space-saving deviceLarge selection of vials and vessels for all analytical problemsVisual and digital status updates
Catalog No. 50-005-80545 Supplier Anton Paar Supplier No. 240857
Please contact your sales representative for a quote or further ordering instructions.
High temperature and pressure for outstanding digestion resultsSoftware-guided procedures make the instrument easy to useIntegrated cooling system for a space-saving deviceLarge selection of vials and vessels for all analytical problemsVisual and digital status updates