Learn More
Thomas Scientific Dry Ice Storage Chest, with 2 spring loaded handles, 3.75 cu. ft., OD: 28.13 x 19.13 x 25.25"; holds up to 200 lb. dry ice pellets
SDP
Catalog No. NC1591254
Encompass_Preferred
Double-wall polyethylene construction filled with urethane foam insulation maintains dry ice quality. Reduces sublimation rates Ideal for dispensing dry ice in a laboratory environment Inner and outer walls are molded of rugged ABS plastic, insulated with 64 mm of expanded polystyrene Lid is hinged at back and has locking hinge on side which can be used to hold lid open Optional Nickel-plated handles and latches
Catalog No. NC1591254 Supplier Thomas Scientific Supplier No. 6391J12
Double-wall polyethylene construction filled with urethane foam insulation maintains dry ice quality. Reduces sublimation rates Ideal for dispensing dry ice in a laboratory environment Inner and outer walls are molded of rugged ABS plastic, insulated with 64 mm of expanded polystyrene Lid is hinged at back and has locking hinge on side which can be used to hold lid open Optional Nickel-plated handles and latches