Learn More
3M™ Heavy Duty Duct Tape
Catalog No. 19070911 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Silver
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-070-911 Black
19-070-912 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-070-911 Supplier 3M™ Supplier No. 7100158377
Add to Cart
Add to Cart
3M Company Heavy Duty Duct Tape is tough tape that resists curling and tears off the roll cleanly for a variety of applications
  • Constructed of polyethylene film laminated to cloth with a synthetic rubber adhesive
  • Designed to provide a formidable, reliable solution for everyday challenges
  • Adheres to numerous substrates, including plastic
  • Sticks immediately and stays on irregular surfaces without lifting
  • Polyethylene, water-resistant backing resists wear and light abrasion
  • Provides a strong, moisture-resistant seal in many containment situations, such as attaching heavy poly draping and pipe sealing
  • Available in black and silver
  • 48 mm x 54.8 m 11 mil
  • 24 individually wrapped roll per case
  • 3M Catalog# DT11 , 3M Catalog# DT11
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polyethylene
Product Type Masking and Duct Tapes
Model DT11
Quantity 24/Pk.