Learn More
MSC Goodway Heavy-Duty Pressure Washer, 1,400 PSI, 1.7 GPM, 115 V
Catalog No. 502836931
Encompass
This heavy-duty pressure washer is a versatile unit suitable for a wide range of traditional pressure washing applications. It can also be converted into a powerful drain cleaner due to its pressure regulator and pulse control, delivering up to 1,400 PSI at 1.7 GPM.
  • Pressure regulator adjusts the PSI to fit the application
  • Pulse control provides additional cleaning force when used as a drain cleaner
  • Heavy-duty construction ensures long life
  • Steel reinforced hose is rugged
  • Stainless steel wand is durable
  • Variable pressure nozzle matches the application
  • Can use hot water for additional cleaning power
  • Locking casters prevent rolling
Catalog No. 50-283-6931 Supplier MSC Supplier No. 60189024
Add to Cart
Add to Cart
Request a Quote
This heavy-duty pressure washer is a versatile unit suitable for a wide range of traditional pressure washing applications. It can also be converted into a powerful drain cleaner due to its pressure regulator and pulse control, delivering up to 1,400 PSI at 1.7 GPM.
  • Pressure regulator adjusts the PSI to fit the application
  • Pulse control provides additional cleaning force when used as a drain cleaner
  • Heavy-duty construction ensures long life
  • Steel reinforced hose is rugged
  • Stainless steel wand is durable
  • Variable pressure nozzle matches the application
  • Can use hot water for additional cleaning power
  • Locking casters prevent rolling