Learn More
Grainger Heavy Duty Recalibratable Gauge
Catalog No. 502751012
Encompass_Preferred
The Haltec Heavy Duty Recalibratable 140X95 is a heavy duty load cell with a 140 ton capacity and a 95 mm diameter. It features a stainless steel construction and hermetically sealed design for protection against harsh environments. The load cell has a 0.05% accuracy with an adjustable zero balance. It can be used for compression or tension applications such as silos, bins, and hoppers. The load cell has a rugged design and is ideal for industrial weighing applications. It requires an excitation voltage of 10V DC and provides an output of 2mV/V. The load cell should be installed properly to ensure optimum performance. Regular calibration is recommended.
Catalog No. 50-275-1012 Supplier Grainger Supplier No. 140X95
The Haltec Heavy Duty Recalibratable 140X95 is a heavy duty load cell with a 140 ton capacity and a 95 mm diameter. It features a stainless steel construction and hermetically sealed design for protection against harsh environments. The load cell has a 0.05% accuracy with an adjustable zero balance. It can be used for compression or tension applications such as silos, bins, and hoppers. The load cell has a rugged design and is ideal for industrial weighing applications. It requires an excitation voltage of 10V DC and provides an output of 2mV/V. The load cell should be installed properly to ensure optimum performance. Regular calibration is recommended.