Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Heavy-Duty SMS Coverall
SDP

Catalog No. 19070133
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-070-133 Small
19-055-365 Medium
19-055-364 Large
19-068-497 X-Large
19-068-498 2X-Large
19-068-499 3X-Large
19-055-366 4X-Large
19-070-134 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-070-133 Supplier Keystone Safety Supplier No. CVLSMSESMWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Keystone Safety Heavy-Duty SMS Coverall is a latex free coverall that is made from high quality materials and comfortable to wear. It conforms to ANSI specs and has a strong/high tensile strength. This durable yet light-weight coverall is disposable with surged seams.

  • Fluid and Tear/Puncture Resistant
  • 100% Latex Free
  • Soft & Durable

Specifications

Product Type Single-use Coveralls
Size Small
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Particle, Liquid
Latex Free Yes
No. of Pockets None
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.