Learn More
Air Science Usa Base Stand for Ductless Fume Hoods, Laminar Flow Cabinets, PCR Workstations, Vented Enclosures, Downflow Workstations, includes Locking Caster Wheels, 24" / 600mm wide units
SDP
Catalog No. NC0984143
Encompass_Preferred
Base Stand, (Metal), Open Frame, Mobile, with Locking Casters, Lower Shelf, Fixed height, 28" Deep. Fits Purair Basic P5-24, DWS24, FLOW24, PCR24, VE24S, Ptefh-24, CA24S, and other 24" / 600mm Nominal Width Models (cart-25)
Catalog No. NC0984143 Supplier Air Science Usa Supplier No. P5CART
Base Stand, (Metal), Open Frame, Mobile, with Locking Casters, Lower Shelf, Fixed height, 28" Deep. Fits Purair Basic P5-24, DWS24, FLOW24, PCR24, VE24S, Ptefh-24, CA24S, and other 24" / 600mm Nominal Width Models (cart-25)