Learn More
Ace Glass, Inc. 30mL Hi-Lube heavy duty stirrer lubrication.
SDP
Catalog No. 509558288
Encompass
30mL container of Hi-Lube heavy duty stirrer lubricant. Lubricant is chlorofluorocarbon grease for use at speeds up to 6000 rpm with ACE standard glass assemblies. High chemical inertness is unaffected by strong acids and alkalis. Soluble in most organic solvents. Suitable for use with oxidizing gases. Has high heat resistance and is thermally stable up to 260 degrees C (500 degrees F). Non-flammable; does not carbonize on decomposition.
Catalog No. 50-955-8288 Supplier Ace Glass, Inc. Supplier No. 811907
30mL container of Hi-Lube heavy duty stirrer lubricant. Lubricant is chlorofluorocarbon grease for use at speeds up to 6000 rpm with ACE standard glass assemblies. High chemical inertness is unaffected by strong acids and alkalis. Soluble in most organic solvents. Suitable for use with oxidizing gases. Has high heat resistance and is thermally stable up to 260 degrees C (500 degrees F). Non-flammable; does not carbonize on decomposition.