Learn More
Cole-Parmer Essentials Heavy Duty Support Stand with 36” S/S Rod
Supplier:  Cole-Parmer 0812625
Encompass_Preferred
Heavy-Duty support base and rod assemblies are made with a cast iron base and a 303 stainless steel rod. The stands base is designed to accommodate vessels up to 12” (30.5 cm) in diameter so to sit close to the support rod. Each of the corners of the four legs base rests on a rubber cushion for added stability. Stand features a corrosion-resistant, electro polished cast iron base.
Catalog No. NC2229356