Learn More
Bachem ([13C6]Leu15)-pTH (1-34) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4072021.1000
Encompass_Preferred
([13C6]Leu15)-pTH (1-34) (human) Trifluoroacetate, H-Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-[13C6]Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OH, 1927927-15-8, C17513C6H291N55O51S2, Mr 4123.71, 1mg. ([13C6]Leu15)-Teriparatide. Heavy isotope-labeled human pTH (1-34) (Teriparatide) useful for pharmacokinetic/pharmacodynamic studies in combination with Bachem product H-4835 pTH (1-34) (human) or the corresponding generic API.
Catalog No. 50-194-6980