Learn More
Heavy Magnesium Oxide, Heavy, Powder, FCC, 96-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| Physical Form | Powder, Needles or Crystals |
| CAS Max % | 100% |
| Quantity | 25 kg |
| Loss on Ignition | 10% |
| Packaging | Poly Bottle |
| Percent Purity | 96 to 100.5% |
| Grade | FCC |
| Chemical Name or Material | Heavy Magnesium Oxide, Heavy, Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.