Learn More
Cytoskeleton, Inc. Heavy Meromyosin Protein HMM, Active F-Actin ATPase, Bovine Cardiac Source, 5 Qty
Catalog No. NC3456423
Encompass
Heavy Meromyosin Protein HMM is a biologically active myosin II fragment sourced from bovine cardiac muscle, essential for studying actin-myosin interactions and ATPase kinetics. This product exhibits F-actin activated ATPase activity and is ideal for biochemical assays investigating enzymatic properties. Supplied as a white lyophilized powder, it supports research into muscle function and contractility, offering a reliable tool for scientists exploring muscle mechanics and pharmacological effects.
Catalog No. NC3456423 Supplier Cytoskeleton, Inc. Supplier No. CSMH03XL
Heavy Meromyosin Protein HMM is a biologically active myosin II fragment sourced from bovine cardiac muscle, essential for studying actin-myosin interactions and ATPase kinetics. This product exhibits F-actin activated ATPase activity and is ideal for biochemical assays investigating enzymatic properties. Supplied as a white lyophilized powder, it supports research into muscle function and contractility, offering a reliable tool for scientists exploring muscle mechanics and pharmacological effects.