Learn More
Perkin Elmer US LLC Heavy Nichrome Wire - Qty. 5
Catalog No. 502111469
Encompass_Preferred
Hangdown wires suspend down from the TGA Furnace Balance Mechanism and catch a sample pan's stirrup and lift the pan from the Autosampler and into the furnace. The hangdown wires come in a variety of materials and kit sizes.
Catalog No. 50-211-1469 Supplier Perkin Elmer US LLC Supplier No. N5370490
Hangdown wires suspend down from the TGA Furnace Balance Mechanism and catch a sample pan's stirrup and lift the pan from the Autosampler and into the furnace. The hangdown wires come in a variety of materials and kit sizes.