Learn More
Sutter Instruments HEAVY POLISHED TUBING W/ FIL.
SDP
Catalog No. NC0319571
Encompass_Preferred
HEAVY POLISHED STANDARD - 1.50mmX0.86mmX10cm. - 250 pieces per packag
Catalog No. NC0319571 Supplier Sutter Instruments Supplier No. FGGBF1508610HP
HEAVY POLISHED STANDARD - 1.50mmX0.86mmX10cm. - 250 pieces per packag