Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, three neck, 1L, 24/40 center, 24/40 sides
SDP
Catalog No. NC0729741
Encompass_Preferred
1L heavy wall, round bottom flask with three vertical necks. Center neck and side necks have 24/40 joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. NC0729741 Supplier Ace Glass, Inc. Supplier No. 6944224
1L heavy wall, round bottom flask with three vertical necks. Center neck and side necks have 24/40 joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.