Learn More
Chemglass Life Sciences Chemglass Heavy Wall Vacuum Tubing, Excelon PVC - 1/4 in ID x 5/8 in OD x 3/16 in wall x 10 ft
SDP
Catalog No. NC2323687
Encompass_Preferred
Heavy-wall Excelon PVC vacuum tubing designed for laboratory and vacuum applications; offers improved vacuum performance, chemical resistance, and sterilization compatibility.
  • Heavy-wall construction provides improved vacuum performance and reduced collapse risk.
  • Dimensions: 1/4 in ID x 5/8 in OD x 3/16 in wall x 10 ft length.
  • Made from PVC for chemical resistance and durability.
  • Temperature range suitable for many lab operations: -58°F to 165°F.
  • Compatible with autoclave and gamma sterilization processes.
  • Meets FDA standards (CFR 21, Part 175.300) for applicable uses.
  • Color transitioning to a blue tint for improved visual identification.
  • Commonly used for laboratory vacuum lines and general tubing applications.
Catalog No. NC2323687 Supplier Chemglass Life Sciences Supplier No. CG735V250
Heavy-wall Excelon PVC vacuum tubing designed for laboratory and vacuum applications; offers improved vacuum performance, chemical resistance, and sterilization compatibility.
  • Heavy-wall construction provides improved vacuum performance and reduced collapse risk.
  • Dimensions: 1/4 in ID x 5/8 in OD x 3/16 in wall x 10 ft length.
  • Made from PVC for chemical resistance and durability.
  • Temperature range suitable for many lab operations: -58°F to 165°F.
  • Compatible with autoclave and gamma sterilization processes.
  • Meets FDA standards (CFR 21, Part 175.300) for applicable uses.
  • Color transitioning to a blue tint for improved visual identification.
  • Commonly used for laboratory vacuum lines and general tubing applications.