Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Heavyweight Polypropylene Coveralls with Hood and Boots

Catalog No. 17184221 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-221 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-184-221 Supplier International Enviroguard™ Supplier No. 2029XL
Add to Cart
Add to Cart

Heavyweight polypropylene coveralls help protect from dirt, grime, and particulates in non-hazardous environments.

  • With attached hood and boots for full body protection
  • Low-cost protection against dirt and grime
  • Made with heavyweight polypropylene fabric for enhanced safety
  • Possible applications include food processing, construction, pharmaceuticals, and environmental cleanup

Specifications

Product Type Light-duty Coveralls
Size X-Large
Gender Unisex
Material Polypropylene (Heavy-duty)
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Latex Free Yes
Model 2029
No. of Pockets None
For Use With (Application) Food Processing, Pharmaceutical, Environmental Cleanup, Construction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.