Learn More
National Safety Apparel™ Heavyweight UltraSoft FR Balaclava
SDP
Catalog No. 17001706 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-706 Color: Navy
1 options
Catalog No. 17-001-706 Supplier National Safety Apparel™ Supplier No. H11RV
Offers durable heavyweight protection while providing the utmost in comfort and flexibility while performing on the job
  • Clothing Type: Hoods
  • Navy
  • Compliance: Cat 3
  • Fabric Weight: 10 oz.
  • Indura ultra soft material
  • One size fits all
Specifications
Size Medium
Material Westex™ UltraSoft™ Rib
Color Navy
Seam Type Stitched
Arc Flash Rating 25 cal/cm²
Certifications/Compliance CAT 3, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462