Learn More
Enzo Life Sciences Hecameg (5g). CAS: 115457-83-5
SDP
Supplier:  Enzo Life Sciences ALX500003G005
Encompass_Preferred
Non-ionic, glucose based very mild detergent for membrane protein purification. Alternative name: 6-O-(N-Heptylcarbamoyl)-methyl-alpha-D-glucopyranoside. Purity: ≥98% (HPLC). Appearance: White to off-white crytalline solid. Solubility: Soluble in water (>100mg/ml at +4°C). Long Term Storage: +4°C.
Catalog No. 50-200-9338