Learn More
Medchemexpress LLC Heclin | 890605-54-6 | 98.2% | 283.32 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11020450MG
Encompass_Preferred
Heclin is an inhibitor of HECT E3 ubiquitin ligases. It inhibits Smurf2, Nedd4, and WWP1 with IC50 values of 6.8, 6.3, and 6.9 μM, respectively, and is suitable for gastric cancer research.
  • HECT E3 ubiquitin ligases inhibitor
  • Inhibits Smurf2, Nedd4, WWP1
  • Can be used for the research of gastric cancer
Catalog No. 50-003-43839