Learn More
Medchemexpress LLC Heclin | 890605-54-6 | 98.2% | 283.32 g/mol | C17H17NO3 | 5 MG
SDP
Catalog No. 5000263407
Encompass_Preferred
Heclin is a small-molecule inhibitor of HECT-family E3 ubiquitin ligases used as a research reagent. It inhibits Smurf2, Nedd4, and WWP1 with micromolar potency and is used in studies of ubiquitin-mediated signaling and cancer biology. Supplied as a powder with recommended cold storage for stability.
  • Inhibits Smurf2, Nedd4, and WWP1 (IC50 ≈ 6-7 μM).
  • High purity (~98.2%) suitable for biochemical assays.
  • Available in small research quantities (5 mg) as a powder.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for extended storage.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-63407 Supplier Medchemexpress LLC Supplier No. HY1102045MG
Heclin is a small-molecule inhibitor of HECT-family E3 ubiquitin ligases used as a research reagent. It inhibits Smurf2, Nedd4, and WWP1 with micromolar potency and is used in studies of ubiquitin-mediated signaling and cancer biology. Supplied as a powder with recommended cold storage for stability.
  • Inhibits Smurf2, Nedd4, and WWP1 (IC50 ≈ 6-7 μM).
  • High purity (~98.2%) suitable for biochemical assays.
  • Available in small research quantities (5 mg) as a powder.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for extended storage.
  • Intended for research use only; not for human or veterinary use.