Learn More
Cayman Chemical Hecogenin Acetate, Purity: ≥98%, CAS: 915-35-5, 10 g
SDP
Catalog No. 502935656
Encompass_Preferred
A steroid sapogenin; reduces the percentage of mobile larvae in goat fecal cultures containing Haemonchus, Oesophagostomum, and Trichostrongylus nematodes (EC50 = 0.49 mg/ml); inhibits development of mechanical hyperalgesia induced by carrageenan, TNF-α, dopamine, and PGE2 at 5, 10, or 20 mg/kg; increases latency to tail withdrawal in the tail flick test in mice at 40 mg/kg
Catalog No. 50-293-5656 Supplier Cayman Chemical Supplier No. 2597710 G
A steroid sapogenin; reduces the percentage of mobile larvae in goat fecal cultures containing Haemonchus, Oesophagostomum, and Trichostrongylus nematodes (EC50 = 0.49 mg/ml); inhibits development of mechanical hyperalgesia induced by carrageenan, TNF-α, dopamine, and PGE2 at 5, 10, or 20 mg/kg; increases latency to tail withdrawal in the tail flick test in mice at 40 mg/kg