Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hederacoside D | 760961-03-3 | MFCD22572710 | 99.3% | 1075.24 | C53H86O22 | 10 MG
SDP

Catalog No. 5000296359
Encompass_Preferred

Hederacoside D is a bioactive triterpenoid saponin isolated from Hedera helix (common ivy). It is provided as a white to off-white solid for research and analytical applications, with high purity and characterized molecular properties. Solubility data indicate excellent solubility in DMSO and formulations suitable for in vivo dosing studies.

  • Bioactive triterpenoid saponin from Hedera helix.
  • High purity (99.3%).
  • Molecular formula C53H86O22 and molecular weight 1075.24.
  • Soluble in DMSO at 100 mg/mL (may require ultrasonic).
  • Supplied in small research pack sizes such as 10 mg.

Catalog No. 50-002-96359 Supplier Medchemexpress LLC Supplier No. HYN025410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hederacoside D is a bioactive triterpenoid saponin isolated from Hedera helix (common ivy). It is provided as a white to off-white solid for research and analytical applications, with high purity and characterized molecular properties. Solubility data indicate excellent solubility in DMSO and formulations suitable for in vivo dosing studies.

  • Bioactive triterpenoid saponin from Hedera helix.
  • High purity (99.3%).
  • Molecular formula C53H86O22 and molecular weight 1075.24.
  • Soluble in DMSO at 100 mg/mL (may require ultrasonic).
  • Supplied in small research pack sizes such as 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.