Learn More
TARGETMOL CHEMICALS INC Hederagenin 28-O-beta-D-glucop
SDP
Catalog No. 502875802
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. Hederagenin-28-beta-D-glucopyranoside, a triterpenoid saponin isolated from Ilex cornuta, protects against H2O2-induced damage in cardiomyocytes. Purity 99.95%
Catalog No. 50-287-5802 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S02085MG
Also available in 1 mL and bulk. Please contact Fisher for quotes. Hederagenin-28-beta-D-glucopyranoside, a triterpenoid saponin isolated from Ilex cornuta, protects against H2O2-induced damage in cardiomyocytes. Purity 99.95%