Learn More
Medchemexpress LLC Hederagenin 28-O-beta-D-glucopyranosyl ester | 53931-25-2 | 5 MG
SDP
Catalog No. 5000420268
Encompass_Preferred
Hederagenin 28-O-beta-D-glucopyranosyl ester is a triterpenoid saponin isolated from Ilex cornuta. It exhibits protective effects against H2O2-induced myocardial cell injury.
  • Triterpenoid saponin
  • Isolated from Ilex cornuta
  • Exhibits protective effects against H2O2-induced myocardial cell injury
  • Store at -20°C, sealed, away from moisture and light
Catalog No. 50-004-20268 Supplier Medchemexpress LLC Supplier No. HYN21905MG
Hederagenin 28-O-beta-D-glucopyranosyl ester is a triterpenoid saponin isolated from Ilex cornuta. It exhibits protective effects against H2O2-induced myocardial cell injury.
  • Triterpenoid saponin
  • Isolated from Ilex cornuta
  • Exhibits protective effects against H2O2-induced myocardial cell injury
  • Store at -20°C, sealed, away from moisture and light