Learn More
Cayman Chemical HederasponIn B 10mg
SDP
Catalog No. 502990395
Encompass_Preferred
A triterpene saponin with antiviral and antioxidant activities; inhibits the cytopathic effect induced by the EV71 genotypes C3 and C4a (EC50s = 24.77 and 41.77 UG/ml, respectively), as well as reduces expression VP2, in Vero cells; inhibits superoxide generation in human neutrophils at 100, 150, and 200 UG/ml
Catalog No. 50-299-0395 Supplier Cayman Chemical Supplier No. 3064110 MG
A triterpene saponin with antiviral and antioxidant activities; inhibits the cytopathic effect induced by the EV71 genotypes C3 and C4a (EC50s = 24.77 and 41.77 UG/ml, respectively), as well as reduces expression VP2, in Vero cells; inhibits superoxide generation in human neutrophils at 100, 150, and 200 UG/ml