Learn More
Cell Signaling Technology Hedgehog Signaling Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502727745
Encompass
The Hedgehog Signaling Antibody Sampler Kit provides an economical means of evaluating key members of the Hedgehog signaling pathway. The kit includes enough antibodies to perform two western blot experiments with each primary antibody.
  • Features antibodies to study canonical Hedgehog ligands, receptors, and signaling transducers
  • Includes eight primary antibodies targeting key pathway proteins
  • Supplied with an HRP-linked anti-rabbit IgG secondary antibody
  • Validated for western blot and immunoprecipitation applications
  • Convenient format for screening multiple pathway members
Catalog No. 50-272-7745 Supplier Cell Signaling Technology Supplier No. 26118T
The Hedgehog Signaling Antibody Sampler Kit provides an economical means of evaluating key members of the Hedgehog signaling pathway. The kit includes enough antibodies to perform two western blot experiments with each primary antibody.
  • Features antibodies to study canonical Hedgehog ligands, receptors, and signaling transducers
  • Includes eight primary antibodies targeting key pathway proteins
  • Supplied with an HRP-linked anti-rabbit IgG secondary antibody
  • Validated for western blot and immunoprecipitation applications
  • Convenient format for screening multiple pathway members