Learn More
Labelmaster Inc HEDPAK 5 Gallon Plastic Container with Outer Carton – Assembled
SDP
Catalog No. NC9712552
Encompass_Preferred
Hedpak5 Gallon The sturdy square-shaped bottle has a molded in grip for easy handling plus 70mm polyethylene Rieke screw cap wi Package comes with a 5-gallon high-density polyethylene bottle and shipping carton
Catalog No. NC9712552 Supplier Labelmaster Inc Supplier No. KH2005
Hedpak5 Gallon The sturdy square-shaped bottle has a molded in grip for easy handling plus 70mm polyethylene Rieke screw cap wi Package comes with a 5-gallon high-density polyethylene bottle and shipping carton