Learn More
Medchemexpress LLC Hee1-2tyr | 2245195-67-7 | 582.67 | C33H30N2O6S | 10 MG
SDP
Catalog No. 5000213409
Encompass_Preferred
HeE1-2Tyr is a research compound described as an RNA-dependent RNA polymerase (RdRp) inhibitor with reported in vitro activity against flaviviruses and coronaviruses. It is supplied as a small-mass powder intended for laboratory research and preparation of concentrated DMSO stock solutions.
  • Inhibits RNA-dependent RNA polymerase activity in vitro.
  • Reported activity against West Nile virus, dengue virus, and SARS-CoV-2.
  • Supplied as a powder suitable for preparing DMSO stock solutions.
  • Compatible with DMSO at 100 MG/ML for concentrated stocks; sonication may be required.
  • Molecular weight 582.67 and CAS number 2245195-67-7 for unambiguous identification.
Catalog No. 50-002-13409 Supplier Medchemexpress LLC Supplier No. HY10074910MG
HeE1-2Tyr is a research compound described as an RNA-dependent RNA polymerase (RdRp) inhibitor with reported in vitro activity against flaviviruses and coronaviruses. It is supplied as a small-mass powder intended for laboratory research and preparation of concentrated DMSO stock solutions.
  • Inhibits RNA-dependent RNA polymerase activity in vitro.
  • Reported activity against West Nile virus, dengue virus, and SARS-CoV-2.
  • Supplied as a powder suitable for preparing DMSO stock solutions.
  • Compatible with DMSO at 100 MG/ML for concentrated stocks; sonication may be required.
  • Molecular weight 582.67 and CAS number 2245195-67-7 for unambiguous identification.