Learn More
Electron Microscopy Sciences Heel Grounder with 1 Mega Ohm Resistor Blue
SDP
Catalog No. 50996257
Encompass_Preferred
E.Z. Close is your low cost logical choice heel grounder. They are made from premium grade non-marking conductive rubber that is highly abrasion and wear resistant. The Velcro fastening system is adjust-able and comfortable. Includes: 18" conductive tab and 1 1/2", two-layer, rubber cup. Built in resistor. Charge decay: less than 0.01 sec.; 2.4x108 ohms . Meets DOD-263A standards (USA).
Catalog No. 50-996-257 Supplier Electron Microscopy Sciences Supplier No. 6061910
E.Z. Close is your low cost logical choice heel grounder. They are made from premium grade non-marking conductive rubber that is highly abrasion and wear resistant. The Velcro fastening system is adjust-able and comfortable. Includes: 18" conductive tab and 1 1/2", two-layer, rubber cup. Built in resistor. Charge decay: less than 0.01 sec.; 2.4x108 ohms . Meets DOD-263A standards (USA).