Learn More
Transforming Technologies Heel Grounder - Stretch Hook and Loop Strap
SDP
Catalog No. 17018492
Change view
Click to view available options
Black
Catalog No.
17-018-492 Black
1 options
Catalog No. 17-018-492 Supplier Transforming Technologies Supplier No. HG1360TT
Add to Cart
Add to Cart
The HG1360 Black ESD Heel Grounders is constructed from durable materials and features a stretch hook and loop closure strap to maximize fit and comfort.
  • MOST POPULAR: Ground mobile personnel with our long-lasting Stretch Hook and Loop ESD heel grounders.
  • LONG LIFE: The reversible cup design allows for longer life. When one side becomes worn, you can flip the strap over and use the other side to extend the life.
  • COMFORT: The stretch Hook and Loop closure strap is made with elastic which creates a comfortable but secure fit around shoe. The elastic strap stretches as you walk which makes for a more comfortable fit.
  • SAFETY: A buried 1meg resistor is built into the strap for safety and to complies with ANSI ESD S2020.
  • OPTIONS: Available in Black. Also available in Blue, Red, and Orange colors as well as Non-Marking Gray Rubber for clean environments.
Specifications
Closure Type Loop
Includes Buried resistor
Length (English) 1.25 in.
Product Type METGUARD Strap
Closure Color Black