Learn More
Grainger Heel Grounding Strap, 1 Megohm
Catalog No. NC0911694
Encompass_Preferred
No-Slip Black Sole, Burgundy Hook And Loop, 1 Megohm Resistor, Moderate Heel Contact Area, ESD SP9.2 Type A, Effectively Grounds Mobile Personnel When They Are In Contact With Conductive And Dissipative Flooring
Catalog No. NC0911694 Supplier Grainger Supplier No. 3XJZ2
No-Slip Black Sole, Burgundy Hook And Loop, 1 Megohm Resistor, Moderate Heel Contact Area, ESD SP9.2 Type A, Effectively Grounds Mobile Personnel When They Are In Contact With Conductive And Dissipative Flooring