Learn More
Grainger SCS Heel Grounding Strap, 1 Pack
Catalog No. NC1454267
Encompass_Preferred
The SCS Heel Grounding Strap is a single pack of heel grounding straps designed to prevent electrostatic discharge. It features a conductive ribbon inside the heel and sole to remove static electricity from the body. The strap has an alligator clip that attaches to the grounding cord. It is made of conductive carbon suffused rubber with a resistance of less than 1.0 megohms. The grounding strap meets ANSI/ESD S20.20 protection standards. It comes in men's sizes 6 through 15.
Catalog No. NC1454267 Supplier Grainger Supplier No. 20FX03
The SCS Heel Grounding Strap is a single pack of heel grounding straps designed to prevent electrostatic discharge. It features a conductive ribbon inside the heel and sole to remove static electricity from the body. The strap has an alligator clip that attaches to the grounding cord. It is made of conductive carbon suffused rubber with a resistance of less than 1.0 megohms. The grounding strap meets ANSI/ESD S20.20 protection standards. It comes in men's sizes 6 through 15.