Learn More
Sklar Instruments Surgi-OR TC Mayo-Hegar Needle Holder, Sklar 5-1/4 in., Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA.
SDP
Catalog No. 501184174 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR TC Mayo-Hegar Needle Holder, Sklar. 5-1/4 in., Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. The Surgi-OR TC Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Tungsten carbide inserts also maximize control and precision. This product is straight, has cross-serrated jaws, and measures 5-1/4 inches overall.
Catalog No. 50-118-4174 Supplier Sklar Instruments Supplier No. 95140
Surgi-OR TC Mayo-Hegar Needle Holder, Sklar. 5-1/4 in., Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. The Surgi-OR TC Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Tungsten carbide inserts also maximize control and precision. This product is straight, has cross-serrated jaws, and measures 5-1/4 inches overall.