Learn More
Lonza Walkersville hEGF - Human Epidermal Growth Factor
Catalog No. NC9165142
Encompass
Epidermal growth factor (EGF), is a growth factor that stimulates cell growth, proliferation, and differentiation by binding to its receptor EGFR. Human EGF is a 6045-Da protein with 53 amino acid residues and three intramolecular disulfide bonds EGF results in cellular proliferation, differentiation, and survival.Epidermal Growth Factor, recombinant is available at a concentration of 3 µg/ml.Content:1 x Vial Containing 3 ug/mL hEGF 0.5 mL
Catalog No. NC9165142 Supplier Lonza Walkersville Supplier No. CC4107
Epidermal growth factor (EGF), is a growth factor that stimulates cell growth, proliferation, and differentiation by binding to its receptor EGFR. Human EGF is a 6045-Da protein with 53 amino acid residues and three intramolecular disulfide bonds EGF results in cellular proliferation, differentiation, and survival.Epidermal Growth Factor, recombinant is available at a concentration of 3 µg/ml.Content:1 x Vial Containing 3 ug/mL hEGF 0.5 mL