Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Hei-PLATE Mix 'n' Heat Magnetic Stirring Hotplate Accessory, Protective Cover
SDP

Catalog No. 13880269 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Hei-PLATE Mix 'n' Heat
Hei-PLATE Mix 20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-880-269 Hei-PLATE Mix 20 L
13-880-270 Hei-PLATE Mix 'n' Heat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-880-269 Supplier Heidolph™ Supplier No. 036306041
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use this Silicone Protective Cover for Hei-PLATE Magnetic Stirrers.

  • Replaceable cover against splashes and dripping water

    Specifications

    Color Silver/White
    For Use With (Equipment) Hei-PLATE Mix 20 L
    Type Silicone Protective Cover
    Material Silicone
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.