Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Hei-PLATE Mix 'n' Heat Magnetic Stirrer Hotplate Accessory, Temperature Sensor
SDP

Catalog No. 13889321 Shop All Heidolph North America Products
Change view
Click to view available options
Model:
Dual PT1000, Stainless Steel
PT1000, Glass Coated
PT1000, Stainless Steel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
13-889-321 PT1000, Glass Coated
13-889-320 PT1000, Stainless Steel
02-707-188 Dual PT1000, Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-889-321 Supplier Heidolph™ Supplier No. 036110361
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Temperature Sensor for measuring and regulating the temperature of the medium, providing precise temperature control.

Single Sensors

  • Choice of stainless steel or glass-coated PT1000
  • 4-pin connection
  • For use with Heidolph Hei-PLATE Expert and Hei-PLATE Ultimate magnetic stirring hotplates

Specifications

Color Silver/White
For Use With (Equipment) Hei-PLATE Mix 'n' Heat Core+, Hei-PLATE Mix 'n' Heat Expert, Hei-PLATE Mix 'n' Heat Ultimate
For Use With (Application) For measuring and regulating the temperature of the medium, providing precise temperature control
Includes Chemical resistant glass coated probe sensor
Model PT1000, Glass Coated
Type Temperature Sensor
Material Glass
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.