Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Hei-SHAKE Centrifuge Tube Attachment with 14 Positions
SDP

Catalog No. 12100030 Shop All Heidolph North America Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
12-100-030 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-100-030 Supplier Heidolph™ Supplier No. 036213736
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Centrifuge tube attachment for Hei-SHAKE Orbital Core platform shaker.

Attachment or rack for holding 14 test tubes such as Falcon 15 mL.

  • Material: stainless steel (frame) / polyoxymethylene (inner part)
  • Characteristics: autoclavable up to 121°C and dishwasher safe; angle adjustment up to 90° attachment without foot part can be used as table frame

This attachment is an optional accessory, must be ordered separately with the base 12-100-026.

Specifications

Autoclavable Autoclavable
Color Silver
Holds Centrifuge Tubes
Material Stainless Steel (Frame), POM (Inner)
Positions 14
Shape Rectangular
Stackable No
Sterility Sterile
For Use With (Equipment) 15 mL centrifuge tubes
Product Type Centrifuge Tube Rack
Shelves 1
Array 7 x 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.